Chalcone 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,3-Diphenyl-2-propen-1-one, Phenyl Styryl ketone, 2-Benzalacetophenone, 2-Benzylideneacetophenone

15943-1000 15943-5000
TS15943-1000EA 62 USD
TS15943-1000 TS15943-5000
Chalcone 97%
Chalcone
Formula: (C₆H₅)C(O)CHCH(C₆H₅)
MW: 208.26 g/mol
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: 53…58 °C
Flash Pt: >112 °C
MDL Number: MFCD00003082
CAS Number: 94-41-7
EINECS: 202-330-2
Merck Index: 13,02045

Order Now

Specification Test Results

Appearance Yellow Granular powder
Infrared spectrum Conforms
Melting point 53 to 58 °C
GC ≥96.0 %
Chloride (Cl) ≤0.01 %
Sulfate (SO4) ≤0.02 %
Iron (Fe) ≤0.002 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR