Acetyl bromide 98%

Supplier: Thermo Scientific Chemicals
Danger

15940-1000 15940-5000
TS15940-1000EA 86 USD
TS15940-1000 TS15940-5000
Acetyl bromide 98%
Acetyl bromide
Formula: C₂H₃BrO
MW: 122.95 g/mol
Boiling Pt: 77 °C (1013 hPa)
Melting Pt: –96 °C
Density: 1.66 g/cm³
Flash Pt: 75 °C
MDL Number: MFCD00000114
CAS Number: 506-96-7
EINECS: 208-061-7
UN: 1716
ADR: 8,II
Merck Index: 13,00084

Order Now

Specification Test Results

Appearance Clear colorless to dark yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4480 to 1.4510 (20°C, 589 nm)
Titration Argentometric ≥97.5 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.01 %
Nitrogen compounds (as N) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR