2-Bromo-2-methylpropane 96% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: tert-Butyl bromide

15926-0025 15926-1000 15926-5000
TS15926-0025EA 1189 USD
TS15926-0025 TS15926-1000 TS15926-5000
2-Bromo-2-methylpropane 96% stabilized
2-Bromo-2-methylpropane
Formula: C₄H₉Br
MW: 137.02 g/mol
Boiling Pt: 72…74 °C (760 mmHg)
Melting Pt: –20 °C
Density: 1.18 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00000125
CAS Number: 507-19-7
EINECS: 208-065-9
UN: 2342
ADR: 3,II
Merck Index: 13,01554

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4269 to 1.4289 (20°C, 589 nm)
GC ≥95 %
Stabilizer potassium carbonate
UV at 280 nm A: ≤0.2
pH 4.0 to 6.0 (1 M in water)
Titration with HClO4 ≥98.5 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.2 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR