1,2-Dibromotetrachloroethane 97%

Supplier: Thermo Scientific Chemicals
Warning

15846-0250 15846-1000
TS15846-0250EA 44 USD
TS15846-0250 TS15846-1000
1,2-Dibromotetrachloroethane 97%
1,2-Dibromotetrachloroethane
Formula: BrCl₂CCBrCl₂
MW: 325.64 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 220…222 °C
Density: 2.71 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000780
CAS Number: 630-25-1
EINECS: 211-136-7

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR