1,12-Dibromododecane 96%

Supplier: Thermo Scientific Chemicals

15845-0250 15845-1000 15845-5000
TS15845-0250EA 82 USD
TS15845-0250 TS15845-1000 TS15845-5000
1,12-Dibromododecane 96%
1,12-Dibromododecane
Formula: BrH₂C(CH₂)₁₀CH₂Br
MW: 328.13 g/mol
Boiling Pt: 215 °C (15 mmHg)
Melting Pt: 37…42 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000226
CAS Number: 3344-70-5
EINECS: 222-096-5

Order Now

Specification Test Results

Appearance Off-white to light brown Crystalline solid
Infrared spectrum Conforms
Melting point 37 to 42 °C
GC ≥95 %
Sulfate (SO4) ≤0.002 %
Calcium (Ca) ≤0.02 %
Iron (Fe) ≤5 ppm
Potassium (K) ≤0.05 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR