2-Chloropyrazine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Chloropyrazine

15806-0100 15806-0500
TS15806-0100EA 75 USD
TS15806-0100 TS15806-0500
2-Chloropyrazine 98%
2-Chloropyrazine
Formula: C₄H₃ClN₂
MW: 114.53 g/mol
Boiling Pt: 153 °C (1013 hPa)
Density: 1.28 g/cm³
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00006124
CAS Number: 14508-49-7
EINECS: 238-517-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5390 (20°C, 589 nm)
Water ≤3 % (K.F.)
GC ≥97.5 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Acidity ≤0.0005 meq/g
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR