p-Toluenesulphonyl hydrazide 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methylbenzenesulphonylhydrazide, 4-Toluenesulphonyl hydrazide, 4-Methylbenzenesulphonhydrazide, 4-Methylbenzenesulphonic acid hydrazide, 4-Methylbenzenesulphonohydrazide, Toluene-4-sulphonohydrazide

15786-0250 15786-1000 15786-5000
TS15786-0250EA 35 USD
TS15786-0250 TS15786-1000 TS15786-5000
p-Toluenesulphonyl hydrazide 97%
p-Toluenesulphonyl hydrazide
Formula: C₇H₁₀N₂O₂S
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 109…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00007588
CAS Number: 1576-35-8
EINECS: 216-407-3
UN: 3226
ADR: 4.1,

Order Now

Specification Test Results

Appearance White to off-white to light grey Powder
Infrared spectrum Conforms
Melting point 105 to 113 °C (Dec)
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR