3-Butyne-1-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxy-1-butyne, But-3-yn-1-ol, 3-Butyn-1-ol, 1-Hydroxy-3-butyne

15733-0050 15733-0250 15733-1000
TS15733-0050EA 51 USD
TS15733-0050 TS15733-0250 TS15733-1000
3-Butyne-1-ol 97%
3-Butyne-1-ol
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 128.8
Melting Pt: -64
Density: 0.926 g/cm³
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00002955
CAS Number: 927-74-2
EINECS: 213-161-9
UN: 1986
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4420 (20°C, 589 nm)
GC ≥96.0 %
Specific optical rotation -10° to -20° (20°C, 589 nm) (c=4 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR