Hexaketocyclohexane octahydrate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Cyclohexanehexone octahydrate, Triquinoyl octahydrate, Cyclohexane-1,2,3,4,5,6-hexone octahydrate

15628-0010 15628-0100
TS15628-0010EA 39 USD
TS15628-0010 TS15628-0100
Hexaketocyclohexane octahydrate 97%
Hexaketocyclohexane octahydrate
Formula: C₆O₆·8H₂O
MW: 312.18 g/mol
Melting Pt: 90…93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149074
CAS Number: 7255-28-9

Order Now

Specification Test Results

Appearance Cream to brown Powder
Infrared spectrum Conforms
Melting point 90 to 103 °C (Dec)
Titration excess NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR