Fumaric acid monoethyl ester 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(E)-4-Ethoxy-4-oxobut-2-enoic acid, Monoethyl fumarate, Ethyl hydrogen fumarate
15620-0250
15620-1000
TS15620-0250EA
68
USD
TS15620-0250
TS15620-1000
Fumaric acid monoethyl ester 95%
Fumaric acid monoethyl ester
Formula:
C₆H₈O₄ MW: 144.13 g/mol Boiling Pt: 147 °C (16 mmHg) Melting Pt: 64…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002699 CAS Number: 2459-05-4 EINECS: 219-544-7 |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 64 to 69 °C |
Titration with NaOH | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...