1,5-Dibromopentane 97%

Supplier: Thermo Scientific Chemicals
Warning

15599-0010 15599-2500
TS15599-0010EA 378 USD
TS15599-0010 TS15599-2500
1,5-Dibromopentane 97%
1,5-Dibromopentane
Formula: BrH₂C(CH₂)₃CH₂Br
MW: 229.94 g/mol
Boiling Pt: 224 °C (1013 hPa)
Melting Pt: –34 °C
Density: 1.68 g/cm³
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00000268
CAS Number: 111-24-0
EINECS: 203-849-7

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5112 to 1.5132 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR