Dibutyl phosphonate P 14.5-16%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Di-n-butylphosphite, Dibutyl phosphite
15506-1000
15506-5000
TS15506-1000EA
38
USD
TS15506-1000
TS15506-5000
Dibutyl phosphonate P 14.5-16%
Dibutyl phosphonate
Formula:
C₈H₁₉O₃P MW: 194.21 g/mol Boiling Pt: 118…119 °C (11 mmHg) Melting Pt: –57 °C Density: 0.995 g/cm³ Flash Pt: 121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066633 CAS Number: 1809-19-4 EINECS: 217-316-1 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4225 to 1.4255 (20°C, 589 nm) |
Assay | 14.5 to 16 % (P) |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...