1,4-Cyclohexanedione 98%

Supplier: Thermo Scientific Chemicals

15483-0100 15483-0500 15483-2500
TS15483-0100EA 38 USD
TS15483-0100 TS15483-0500 TS15483-2500
1,4-Cyclohexanedione 98%
1,4-Cyclohexanedione
Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 227
Melting Pt: 76…80 °C
Flash Pt: 132 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001606
CAS Number: 637-88-7
EINECS: 211-306-0

Order Now

Specification Test Results

Appearance Yellow to beige Powder or flakes
Infrared spectrum Conforms
Melting point 76 to 80 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR