4-Cyanobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

15455-0050 15455-0250
TS15455-0050EA 86 USD
TS15455-0050 TS15455-0250
4-Cyanobenzoyl chloride 98%
4-Cyanobenzoyl chloride
Formula: C₈H₄ClNO
MW: 165.58 g/mol
Boiling Pt: 150…151 °C (25 mmHg)
Melting Pt: 66…70 °C
MDL Number: MFCD00001822
CAS Number: 6068-72-0
EINECS: 228-005-5
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 66 to 70 °C
Titration RCOCl ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR