3-Bromoquinoline 98%

Supplier: Thermo Scientific Chemicals
Warning

15427-0250 15427-1000
TS15427-0250EA 96 USD
TS15427-0250 TS15427-1000
3-Bromoquinoline 98%
3-Bromoquinoline
Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 274 °C (1013 hPa)
Melting Pt: 13…15 °C
Density: 1.53 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00006767
CAS Number: 5332-24-1
EINECS: 226-237-1
UN: 2810

Order Now

Specification Test Results

Appearance Clear colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.6630 to 1.6660 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR