1-Bromo-2-methylnaphthalene 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

15426-0250 15426-1000
TS15426-0050EA 32.96 USD
TS15426-0050 TS15426-0250 TS15426-1000
1-Bromo-2-methylnaphthalene 90%, Technical Grade
1-Bromo-2-methylnaphthalene
Formula: C₁₁H₉Br
MW: 221.1 g/mol
Boiling Pt: 296 °C (760 mmHg)
Density: 1.418 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00003871
CAS Number: 2586-62-1
EINECS: 219-966-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.6450 to 1.6530 (20°C, 589 nm)
GC ≥88 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR