3-Toluoyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Methylbenzoyl chloride, m-Methylbenzoyl chloride, m-Toluoyl chloride, 3-Methylbenzene-1-carbonyl chloride

15341-0050 15341-1000 15341-5000
TS15341-0050EA 33 USD
TS15341-0050 TS15341-1000 TS15341-5000
3-Toluoyl chloride 99%
3-Toluoyl chloride
Formula: C₈H₇ClO
Boiling Pt: 86 °C (5 mmHg)
Melting Pt: –23 °C
Density: 1.17 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00000681
CAS Number: 1711-06-4
EINECS: 216-976-8
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5475 to 1.5495 (20°C, 589 nm)
Titration RCOCl ≥98.5 %
UV at 280 nm A: ≤0.050
pH 10.9 to 11.6 (1 % in water at 25°C)
Titration with HClO4 97.5 to 102.5 % (on dry basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR