2-Toluoyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Methylbenzoyl chloride, o-Methylbenzoyl chloride, o-Toluoyl chloride, 2-Methylbenzene-1-carbonyl chloride

15337-0050 15337-1000 15337-5000
TS15337-0050EA 31 USD
TS15337-0050 TS15337-1000 TS15337-5000
2-Toluoyl chloride 99%
2-Toluoyl chloride
Formula: C₈H₇ClO
Boiling Pt: 88…90 °C (12 mmHg)
Melting Pt: –18 °C
Density: 1.18 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00000668
CAS Number: 933-88-0
EINECS: 213-273-8
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow to pink Liquid
Infrared spectrum Conforms
Refractive index 1.5540 to 1.5560 (20°C, 589 nm)
Titration RCOCl ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR