3,3-Dimethylpentane 99%

Supplier: Thermo Scientific Chemicals
Danger

15266-0010 15266-0050 15266-0250
TS15266-0010EA 48 USD
TS15266-0010 TS15266-0050 TS15266-0250
3,3-Dimethylpentane 99%
3,3-Dimethylpentane
Formula: C₇H₁₆
Boiling Pt: 86 °C (760 mmHg)
Melting Pt: –135 °C
Density: 0.69 g/cm³
Flash Pt: –6 °C
Storage Temperature: Ambient
MDL Number: MFCD00009322
CAS Number: 562-49-2
EINECS: 209-230-8
UN: 1206
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3900 to 1.3920 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR