2-Methyl-1,3-cyclopentanedione 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Methylcyclopentane-1,3-dione

15232-0250 15232-1000
TS15232-0250EA 118 USD
TS15232-0250 TS15232-1000
2-Methyl-1,3-cyclopentanedione 98%
2-Methyl-1,3-cyclopentanedione
Formula: C₆H₈O₂
MW: 112.13 g/mol
Melting Pt: 212…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00001406
CAS Number: 765-69-5
EINECS: 212-153-2

Order Now

Specification Test Results

Appearance Light beige to tan Crystalline powder
Infrared spectrum Conforms
Melting point 212 to 216 °C
GC ≥97.5 %
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR