4-Nitrophenethyl bromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(4-Nitrophenyl)-1-bromoethane, 1-(2-Bromoethyl)-4-nitrobenzene
15162-0050
15162-0250
TS15162-0050EA
47
USD
TS15162-0050
TS15162-0250
4-Nitrophenethyl bromide 98%
4-Nitrophenethyl bromide
Formula:
C₈H₈BrNO₂ MW: 230.06 g/mol Melting Pt: 67…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007386 CAS Number: 5339-26-4 EINECS: 226-271-7 |
Specification Test Results
Appearance | Yellow Adhering crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 67 to 70 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...