(S)-2-Hydroxy-3-phenylpropanoic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: L-β-Phenyllactic acid, (S)-(-)-3-Phenyllactic Acid, L-3-Phenyllactic Acid, L(-)-3-Phenyllactic acid, (S)-(-)-2-Hydroxy-3-phenylpropionic Acid, L-(-)-3-Phenyllactic acid

15092-0010 15092-0050 15092-0250
TS15092-0010EA 33 USD
TS15092-0010 TS15092-0050 TS15092-0250
(S)-2-Hydroxy-3-phenylpropanoic acid 98%
(S)-2-Hydroxy-3-phenylpropanoic acid
Formula: C₉H₁₀O₃
MW: 166.17 g/mol
Melting Pt: 121…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004244
CAS Number: 20312-36-1

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or needles
Infrared spectrum Conforms
Melting point 121 to 125 °C
TLC ≥97.5 %
Specific optical rotation -17° to -20° (24 °C, 589 nm) (c=1, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR