Allylthiourea 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Allylthiourea, Tiosinamine, 1-Allyl-2-thiourea, Allyl thiourea, Rhodalline, Thiosinamine, N-Allylthiourea
14880-0250
14880-1000
14880-5000
TS14880-0250EA
40
USD
TS14880-0250
TS14880-1000
TS14880-5000
Allylthiourea 98%
Allylthiourea
Formula:
H₂CCHCH₂NHC(S)NH₂ MW: 116.19 g/mol Boiling Pt: 191 °C (1013 hPa) Melting Pt: 70…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004940 CAS Number: 109-57-9 EINECS: 203-683-5 UN: 2811 ADR: 6.1,III Merck Index: 12,09500 |
Specification Test Results
Appearance | White to light beige Powder or crystals or granules and/or chunks |
Infrared spectrum | Conforms |
Melting point | 70 to 78 °C |
Loss on drying | ≤1.5 % (50 °C, 2 hrs) |
Titration with HClO4 | 97.5 to 102.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...