4-Bromobenzenesulphonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

14876-0250 14876-1000
TS14876-0250EA 60 USD
TS14876-0250 TS14876-1000
4-Bromobenzenesulphonyl chloride 98%
4-Bromobenzenesulphonyl chloride
Formula: C₆H₄BrClO₂S
MW: 255.52 g/mol
Boiling Pt: 153 °C (15 mmHg)
Melting Pt: 74…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00007437
CAS Number: 98-58-8
EINECS: 202-683-2
UN: 3261
ADR: 8,II
Merck Index: 13,01391

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 74 to 77 °C
Titration Argentometric ≥97.5 % (Ex CL)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR