Benzenesulphonyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

14848-0010 14848-2500
TS14848-0010EA 202 USD
TS14848-0010 TS14848-2500
Benzenesulphonyl chloride 99%
Benzenesulphonyl chloride
Formula: C₆H₅ClO₂S
Boiling Pt: 251…252 °C (760 mmHg)
Melting Pt: 14.5 °C
Density: 1.384 g/cm³
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00007426
CAS Number: 98-09-9
EINECS: 202-636-6
UN: 2225
ADR: 8,III
Merck Index: 13,01072

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5530 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR