1,2-Dibromo-1,2-diphenylethane 96%

Supplier: Thermo Scientific Chemicals
Danger

14795-0050 14795-0250
TS14795-0050EA 84 USD
TS14795-0050 TS14795-0250
1,2-Dibromo-1,2-diphenylethane 96%
1,2-Dibromo-1,2-diphenylethane
Formula: C₆H₅(CHBr)₂C₆H₅
MW: 340.06 g/mol
Melting Pt: 235…241 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00000137
CAS Number: 5789-30-0
EINECS: 227-324-7
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5071ce04f1320e70080fd2e545601e6c65d01f23&vid=1842fa1a5f1421863878c2f18cd43a3e8565af79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to yellow to light orange Powder
Infrared spectrum Conforms
Melting point 234 to 241 °C
HPLC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR