1,1,2,2-Tetrachloroethane 98.5%

Supplier: Thermo Scientific Chemicals
Danger

14794-0025 14794-0010 14794-0250 14794-1000
TS14794-0025EA 399 USD
TS14794-0025 TS14794-0010 TS14794-0250 TS14794-1000
1,1,2,2-Tetrachloroethane 98.5%
1,1,2,2-Tetrachloroethane
Formula: Cl₂HCCHCl₂
MW: 167.85 g/mol
Boiling Pt: 147 °C (760 mmHg)
Melting Pt: –43 °C
Density: 1.58 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000848
CAS Number: 79-34-5
EINECS: 201-197-8
UN: 1702
ADR: 6.1,II
Merck Index: 13,09264
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ad9914010b65d5d21f26f85ccb3fc00111fc8b3&vid=da3549d1364274a05dcc36a80f09745b568776d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4930 to 1.4950 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR