3-Bromothiophene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Thienyl bromide

14780-0250 14780-1000 14780-5000
TS14780-0250EA 104 USD
TS14780-0250 TS14780-1000 TS14780-5000
3-Bromothiophene 97%
3-Bromothiophene
Formula: C₄H₃BrS
MW: 163.04 g/mol
Boiling Pt: 158 °C (760 mmHg)
Density: 1.74 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00005464
CAS Number: 872-31-1
EINECS: 212-821-3
UN: 3384
ADR: 6.1,

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5905 to 1.5925 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR