3-bromofuran-2,5-dione 97%

Supplier: Thermo Scientific Chemicals
Danger

14745-0100 14745-0500
TS14745-0100EA 80 USD
TS14745-0100 TS14745-0500
3-bromofuran-2,5-dione 97%
3-bromofuran-2,5-dione
Formula: C₄HBrO₃
Boiling Pt: 215 °C (760 mmHg)
Density: 1.905 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00005519
CAS Number: 5926-51-2
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5410 to 1.5430 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR