Pentafluorobenzenesulphonyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3,4,5,6-Pentafluorobenzenesulphonyl chloride, Pentafluorobenzene-1-sulphonyl chloride

14711-0050 14711-0250
TS14711-0010EA 38.11 USD
TS14711-0010 TS14711-0050 TS14711-0250
Pentafluorobenzenesulphonyl chloride 99%
Pentafluorobenzenesulphonyl chloride
Formula: C₆ClF₅O₂S
MW: 266.58 g/mol
Boiling Pt: 211 °C (760 mmHg)
Density: 1.796 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007427
CAS Number: 832-53-1
EINECS: 212-620-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4780 to 1.4800 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR