1-Ethoxy-2,2,2-trifluoroethanol 90%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trifluoroacetaldehyde ethyl hemiacetal, Trifluoroacetaldehydeethylhemiacetal

13971-0050 13971-0250
TS13971-0050EA 70 USD
TS13971-0050 TS13971-0250
1-Ethoxy-2,2,2-trifluoroethanol 90%
1-Ethoxy-2,2,2-trifluoroethanol
Formula: C₄H₇F₃O₂
MW: 144.09 g/mol
Boiling Pt: 104…105 °C (745 mmHg)
Flash Pt: 39 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000441
CAS Number: 433-27-2
EINECS: 207-087-6
UN: 1989
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3410 to 1.3440 (20°C, 589 nm)
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR