Triethyl phosphonoacetate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl 2-(diethoxyphosphoryl)acetate, Diethyl [(ethoxycarbonyl)methyl]phosphonate

13970-0025 13970-1000 13970-5000
TS13970-0025EA 1138 USD
TS13970-0025 TS13970-1000 TS13970-5000
Triethyl phosphonoacetate 97%
Triethyl phosphonoacetate
Formula: C₈H₁₇O₅P
Boiling Pt: 260…262 °C (760 mmHg)
Melting Pt: –24 °C
Density: 1.13 g/cm³
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00009177
CAS Number: 867-13-0
EINECS: 212-757-6

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4320 (20°C, 589 nm)
GC ≥96.0 %
Color scale ≤50 APHA
Acid value ≤2 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR