2,4,6-Trichloropyrimidine 99%

Supplier: Thermo Scientific Chemicals
Warning

13949-0250 13949-1000
TS13949-0250EA 89 USD
TS13949-0250 TS13949-1000
2,4,6-Trichloropyrimidine 99%
2,4,6-Trichloropyrimidine
Formula: C₄HCl₃N₂
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 23…25 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00006063
CAS Number: 3764-01-0
EINECS: 223-183-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5690 to 1.5710 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR