p-Toluenesulphinic acid sodium salt hydrate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
13901-1000
13901-5000
TS13901-1000EA
49
USD
TS13901-1000
TS13901-5000
p-Toluenesulphinic acid sodium salt hydrate 98+%
p-Toluenesulfinic acid sodium salt hydrate
Formula:
C₇H₇NaO₂S .xH₂O Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149640 CAS Number: 207801-20-5 |
Specification Test Results
Appearance | White to off-white Crystalline powder or flakes |
Infrared spectrum | Conforms |
Water | ≤5 % (K.F.) |
Titration with HClO4 | ≥98.0 % (On dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...