Tetramethylene sulphoxide 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Tetrahydrothiophene 1-oxide

13844-0050 13844-0250 13844-1000
TS13844-0050EA 33 USD
TS13844-0050 TS13844-0250 TS13844-1000
Tetramethylene sulphoxide 97%
Tetramethylene sulphoxide
Formula: C₄H₈OS
Flash Pt: >112 °C
MDL Number: MFCD00005477
CAS Number: 1600-44-8
EINECS: 216-493-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5190 to 1.5230 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR