3-Iodopyridine 99%

Supplier: Thermo Scientific Chemicals
Danger

13744-0010 13744-0100
TS13744-0010EA 37 USD
TS13744-0010 TS13744-0100
3-Iodopyridine 99%
3-Iodopyridine
Formula: C₅H₄IN
Boiling Pt: 90…92 °C (14 mmHg)
Melting Pt: 53…56 °C
Flash Pt: 106 °C
MDL Number: MFCD00023553
CAS Number: 1120-90-7
EINECS: 214-322-6

Order Now

Specification Test Results

Appearance Light beige Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 56 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR