4-Cyano-4-phenylcyclohexanone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Oxo-1-phenylcyclohexanecarbonitrile, 4-Cyano-4-phenylcyclohexanone

13490-0050 13490-0250
TS13490-0050EA 39 USD
TS13490-0050 TS13490-0250
4-Cyano-4-phenylcyclohexanone 97%
4-Cyano-4-phenylcyclohexanone
Formula: C₁₃H₁₃NO
MW: 199.24
Melting Pt: 114…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00044816
CAS Number: 25115-74-6
EINECS: 246-631-7
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to beige Crystalline solid
Infrared spectrum Conforms
Melting point 114 to 119 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR