6-Methylindole 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Methyl-1H-indole

13388-0010 13388-0050 13388-2500
TS13388-0010EA 196 USD
TS13388-0010 TS13388-0050 TS13388-2500
6-Methylindole 99%
6-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 112 °C (5 mmHg)
Melting Pt: 30…31 °C
Density: 1.05 g/cm³
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005682
CAS Number: 3420-02-8
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear yellow to light brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.6060 to 1.6080 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR