(±)-2-Piperazinecarboxylic acid dihydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Piperazine-2-carboxylic acid dihydrochloride

13366-0100 13366-0500
TS13366-0100EA 83 USD
TS13366-0100 TS13366-0500
(±)-2-Piperazinecarboxylic acid dihydrochloride 98%
(±)-2-Piperazinecarboxylic acid dihydrochloride
Formula: C₅H₁₂Cl₂N₂O₂
MW: 203.07 g/mol
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00044803
CAS Number: 3022-15-9

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 %
Titration with HClO4 97.5 to 102.5 %
NMR (31P-NMR) Consistent with structure
Specific optical rotation +35° to +45° (20°C, 589 nm) (c=1 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR