2-Carboxyphenyl salicylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Salicyloyloxybenzoic Acid, O-Salicyloylsalicylic Acid, Salicylic Acid 2-Carboxyphenyl Ester, Salsalate

13340-0250 13340-1000
TS13340-0250EA 75 USD
TS13340-0250 TS13340-1000
2-Carboxyphenyl salicylate 98%
2-Carboxyphenyl salicylate
Formula: C₁₄H₁₀O₅
MW: 258.23 g/mol
Melting Pt: 139…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00020252
CAS Number: 552-94-3

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %
NMR (1H-NMR) Consistent with structure
NMR (13C-NMR) Consistent with structure
NMR (31P-NMR) Consistent with structure
Elemental Analysis 76.05 to 82.40 % C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR