Succinyl chloride ca. 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Succinyl dichloride

13284-0250 13284-1000 13284-5000
TS13284-0250EA 46 USD
TS13284-0250 TS13284-1000 TS13284-5000
Succinyl chloride ca. 95%
Succinyl chloride
Formula: C₄H₄Cl₂O₂
MW: 154.98 g/mol
Boiling Pt: 190 °C (760 mmHg)
Melting Pt: 16…17 °C
Density: 1.407 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00000749
CAS Number: 543-20-4
EINECS: 208-838-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4670 to 1.4720 (20°C, 589 nm)
Titration RCOCl ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR