Propiolic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acetylenecarboxylic Acid, 2-Propynoic acid
13150-0100
13150-0250
13150-1000
TS13150-0100EA
89
USD
TS13150-0100
TS13150-0250
TS13150-1000
Propiolic acid 98%
Propiolic acid
Formula:
C₃H₂O₂ Boiling Pt: 153…157 °C (760 mmHg) Melting Pt: 16 °C Density: 1.13 g/cm³ Flash Pt: 58 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004360 CAS Number: 471-25-0 EINECS: 207-437-8 UN: 2920 ADR: 8,II Merck Index: 12,08006 |
Specification Test Results
Appearance | Clear colorless to brown Liquid or solid |
Infrared spectrum | Conforms |
Refractive index | 1,4305 to 1,4345 (20 °C, 589 nm) |
Titration with NaOH | 97,5 to 102,5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...