Propagyl amine hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Propargylamine hydrochloride, Prop-2-yn-1-amine hydrochloride

13147-0100 13147-0010
TS13147-0100EA 208 USD
TS13147-0100 TS13147-0010
Propagyl amine hydrochloride 95%
Propagyl amine hydrochloride
Formula: C₃H₆ClN
MW: 91.54 g/mol
Melting Pt: 175…182 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012907
CAS Number: 15430-52-1
EINECS: 239-440-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to beige to brown Crystalline powder
Infrared spectrum Conforms
Melting point 175 to 182 °C
Titration Argentometric 94 to 106 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR