Diphenyl sulphoxide 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Phenyl sulphoxide, sulphinyldibenzene
13097-0250
13097-1000
TS13097-0250EA
58
USD
TS13097-0250
TS13097-1000
Diphenyl sulphoxide 97%
Diphenyl sulphoxide
Formula:
C₁₂H₁₀OS MW: 202.28 g/mol Boiling Pt: 206…208 °C (13 mmHg) Melting Pt: 66…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002085 CAS Number: 945-51-7 EINECS: 213-415-9 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 66 to 71 °C |
Water | ≤0.5 % (K.F.) |
GC | ≥96.0 % |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...