Diphenyl sulphide 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diphenylsulphane

13095-0250 13095-1000
TS13095-0250EA 47 USD
TS13095-0250 TS13095-1000
Diphenyl sulphide 99%
Diphenyl sulphide
Formula: C₁₂H₁₀S
MW: 186.28 g/mol
Boiling Pt: 296
Melting Pt: -40
Density: 1.11 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003064
CAS Number: 139-66-2
EINECS: 205-371-4
Merck Index: 12,07467

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6317 to 1.6337 (20°C, 589 nm)
GC ≥98.5 %
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR