Phenylacetyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
13030-1000
13030-5000
TS13030-1000EA
53
USD
TS13030-1000
TS13030-5000
Phenylacetyl chloride 98%
Phenylacetyl chloride
Formula:
C₈H₇ClO Boiling Pt: 94…95 °C (12 mmHg) Melting Pt: 91 °C Density: 1.16 g/cm³ Flash Pt: 102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000729 CAS Number: 103-80-0 EINECS: 203-146-5 UN: 2577 ADR: 8,II |
Specification Test Results
Appearance | Clear light yellow to pink-red Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5320 to 1.5330 (20°C, 589 nm) |
GC | ≥97.5 % |
Titration RCOCl | ≥97.5 % |
Titration RCOOH | ≤2 % |
Free acid (HCl) | Traces |
Color scale | ≤20 APHA |
Residue after evaporation | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...