3,4,9,10-Perylenetetracarboxylic dianhydride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Anthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone, Pigment Red 224, PTCDA

13008-0250 13008-1000
TS13008-0250EA 56 USD
TS13008-0250 TS13008-1000
3,4,9,10-Perylenetetracarboxylic dianhydride 98%
3,4,9,10-Perylenetetracarboxylic dianhydride
Formula: C₂₄H₈O₆
MW: 392.32 g/mol
Melting Pt: >350 °C
Storage Temperature: Ambient
MDL Number: MFCD00006916
CAS Number: 128-69-8

Order Now

Specification Test Results

Appearance Dark red Powder
Infrared spectrum Conforms
Assay ≥97.5 %
Water ≤1 % (K.F.)
Stabilizer 150 to 400 ppm (BHT)
Peroxides ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR