n-Octadecane 99+%

Supplier: Thermo Scientific Chemicals
Danger

12929-0010 12929-0050 12929-2500
TS12929-0010EA 339 USD
TS12929-0010 TS12929-0050 TS12929-2500
n-Octadecane 99+%
n-Octadecane
Formula: H₃C(CH₂)₁₆CH₃
Boiling Pt: 317 °C (760 mmHg)
Melting Pt: 28…31 °C
Density: 0.777 g/cm³
Flash Pt: 166 °C
Storage Temperature: Ambient
MDL Number: MFCD00009007
CAS Number: 593-45-3
EINECS: 209-790-3

Order Now

Specification Test Results

Appearance White to yellow Solid or liquid
Infrared spectrum Conforms
Melting point 28 to 31 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR