n-Octacosane 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: N-octacosane

12927-0050 12927-0250 12927-1000 12927-5000
TS12927-0050EA 21 USD
TS12927-0050 TS12927-0250 TS12927-1000 TS12927-5000
n-Octacosane 99+%
n-Octacosane
Formula: H₃C(CH₂)₂₆CH₃
MW: 394.77 g/mol
Boiling Pt: 440 °C (760 mmHg)
Melting Pt: 61…64 °C
Density: 0.8067 g/cm³
Flash Pt: 227 °C
Storage Temperature: Ambient
MDL Number: MFCD00009355
CAS Number: 630-02-4
EINECS: 211-125-7

Order Now

Specification Test Results

Appearance White Glistening powder or flakes
Infrared spectrum Conforms
Melting point 61 to 64 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR