1-Methyl-2-thiourea 97%

Supplier: Thermo Scientific Chemicals
Danger

12790-0250 12790-1000
TS12790-0250EA 50 USD
TS12790-0250 TS12790-1000
1-Methyl-2-thiourea 97%
1-Methyl-2-thiourea
Formula: C₂H₆N₂S
MW: 90.15 g/mol
Melting Pt: 118…123 °C
MDL Number: MFCD00004938
CAS Number: 598-52-7
EINECS: 209-936-6
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 118 to 123 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR